Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Neck Cape Fastener Kit
SDP

Catalog No. 18444050
Change view
Click to view available options
Fastener Kit; Color: Black; Material: Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-050 Fastener Kit; Color: Black; Material: Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-050 Supplier Gentex™ Supplier No. PR02065SP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PureFlo ESM+ neck cape fastener kit includes screws (12), fastener studs, washers, and nuts.

  • Accessory for PureFlo ESM+ Neck Cape

Specifications

Description Neck Cape Fastener Kit
Color Black
Disposable Single-use
Material Plastic
Product Type Fastner Kit
Size One Size
For Use With (Equipment) PureFlo ESM+
Includes Screws (12), fastener studs, washers, nuts
Latex Free Yes
Model PR02065SP
Quantity 12/Pk.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.