Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ PF60 Welding PAPR
SDP

Catalog No. 18444069
Change view
Click to view available options
Welding PAPR; Model: PR05814-B-2A1; Includes: Black Stedprene FR Neck Cape, HE Filter, ADF Welding Lens, Internal and External Switches, Black Hard Hat
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-069 Welding PAPR; Model: PR05814-B-2A1; Includes: Black Stedprene FR Neck Cape, HE Filter, ADF Welding Lens, Internal and External Switches, Black Hard Hat
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-069 Supplier Gentex™ Supplier No. PR05801V2A1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Head mounted self-contained PAPR ideal for welding, fabrication, and radiant heat hazards.

  • No waist belts, hoses, or cables for maximum mobility
  • High 8.1 CFM airflow rate
  • Cinched Stedprene fire-resistant drawstring neck cape
  • Hard Hat provides ANSI Z89.1 compliant head protection
  • Choose from black or hi vis color options
  • Quick release adjustable headband for optimal balance and fit
  • Enhanced IP67 sealed blower/motor assembly

Specifications

ANSI Standard Head Protection: ANSI Z89.1 Type 1 Class G; Face Shield/Eye Protection ANSI Z87.1+: 2015 Certified; SEI Certified and Listed
Assigned Protection Factor (APF) 1000
Battery Life 7 hours with two batteries
Battery Type 5 Cell Nickel Metal Hydride
Cartridge/Filter PARTICULATE
Certifications (NIOSH) NIOSH Approved
Product Type PAPR System for Welding
Size One Size
Filter Efficiency HEPA (HE) or HEPA/HF/HC *Nuisance OV
Filter Efficiency (NIOSH) HEPA
Flow Rate 8.1 CFM or 230 L/min.
Includes PAPR, Battery and Charger
Model PR05801-V-2A1
Noise Level Under 70 dB
Quantity 1 Ea.
Recharge Time 2.5 hr.
Replaceable Filter One Size
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.