Learn More
Gentex™ PureFlo ESM+ Power Source Replacement Parts and Accessories

Description
- Power source replacement parts and accessories for the PureFlo ESM+ system include
- Battery pack
- Battery charger
- Battery multicharger - 16 bank
Specifications
Specifications
| Color | Black |
| Disposable | Reusable |
| Product Type | Battery Pack |
| Size | One Size |
| For Use With (Equipment) | PureFlo ESM+ PAPR |
| Latex Free | Yes |
| Model | PR05806SP |
| Quantity | 1 Ea. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.