Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Premium Head Lining Kit
SDP

Catalog No. 18444056
Change view
Click to view available options
Head Lining Kit; Includes: 1 Brow Pad, 2 Nape Pads; Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-056 Head Lining Kit; Includes: 1 Brow Pad, 2 Nape Pads; Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-056 Supplier Gentex™ Supplier No. PR02605SP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Premium head lining kit (1 brow pad and 2 nape pads) for use with the PureFlo ESM+ PAPR.

  • Premium head lining kit
  • 1 Brow Pad And 2 Nape Pads
  • For use with the PureFlo ESM+ PAPR.

Specifications

Description Premium head lining kit
Color Black
Disposable Single-use
Material Cotton
Product Type Comfort Pad
Size One Size
For Use With (Equipment) PureFlo ESM+ PAPR
Includes 1 Brow Pad, 2 Nape Pads
Latex Free Yes
Model PR02605SP
Quantity 1 Ea.
Sterility Non-sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.