Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Visor Assembly
SDP

Catalog No. 18444054
Change view
Click to view available options
Model:
PR02050SP
PR02480SP
Quantity:
1 Ea.
4/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Quantity
18-444-054 PR02480SP 1 Ea.
18-444-055 PR02050SP 4/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-444-054 Supplier Gentex™ Supplier No. PR02480SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Spare visor assembly (Non Welding Unit) for use with the PureFlo ESM+ PAPR.

  • Spare visor assembly (Non Welding Unit)
  • For use with the PureFlo ESM+ PAPR
  • Color: Clear

Specifications

Description Spare visor assembly (Non Welding Unit) for use with the PureFlo ESM+ PAPR.
Color Clear
Disposable Reusable
Product Type Visor
Size One Size
For Use With (Equipment) PureFlo PF60 ESM+
Latex Free Yes
Model PR02480SP
Quantity 1 Ea.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.