Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Visor Protector
SDP

Catalog No. 17010064
Change view
Click to view available options
Quantity:
1 Ea.
4/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-010-064 1 Ea.
17-010-065 4/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 17-010-064 Supplier Gentex™ Supplier No. PR01545SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Visor protector fits over the standard PureFlo ESM and ESM+ PAPR visor and protects from scratches and harsh environments.

  • Spare visor protector for PureFlo ESM and ESM+ PAPR systems
  • Accessory

Specifications

Color Clear
Disposable Single-use
Material Acetate
Product Type Visor Protector
For Use With (Equipment) PureFlo ESM and ESM+ PAPR Systems
Model PR01545SP
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.