Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo ESM+ Visor Protector
SDP

Catalog No. p-7227704
Click to view available options
Quantity:
1 Ea.
4/Pk.
This item is not returnable. View return policy
This item is not returnable. View return policy

Visor protector fits over the standard PureFlo ESM and ESM+ PAPR visor and protects from scratches and harsh environments.

  • Spare visor protector for PureFlo ESM and ESM+ PAPR systems
  • Accessory

Specifications

Color Clear
Disposable Single-use
Material Acetate
Product Type Visor Protector
For Use With (Equipment) PureFlo ESM and ESM+ PAPR Systems
Model PR01545SP
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.