Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo PF50 ESM+ Bump Cap
SDP

Catalog No. 18444051
Change view
Click to view available options
Bump Cap; For Use With: PureFlo PF50 ESM+; Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-051 Bump Cap; For Use With: PureFlo PF50 ESM+; Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-051 Supplier Gentex™ Supplier No. PR05504SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Bump Cap for PureFlo PF50 ESM+ PAPR.

  • Accessory for PureFlo PF50 ESM+ PAPR

Specifications

Description Bump Cap for PureFlo PF50 ESM+ PAPR
Color Gray
Material Plastic
Product Type Bump Cap Assembly
Size One Size
For Use With (Equipment) PureFlo PF50 ESM+
Latex Free Yes
Model PR05504SP
Quantity 1 Ea.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.