Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gentex™ PureFlo PF50 ESM+ Visor Assembly
SDP

Catalog No. 18444057
Change view
Click to view available options
Visor Assembly; For Use With: PureFlo PF50 ESM+; Color: Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
18-444-057 Visor Assembly; For Use With: PureFlo PF50 ESM+; Color: Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-444-057 Supplier Gentex™ Supplier No. PR05502SP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

4 pack visor assembly replacement (P Series) for use with the PureFlo PF50 ESM+ PAPR.

  • Accessory for PureFlo PF50 ESM+ PAPR

Specifications

Description Visor Assembly for PureFlo P-Series PAPR
Color Clear
Disposable Single-use
Product Type Visor Assembly
Size One Size
For Use With (Equipment) PureFlo PF50 ESM+
Latex Free Yes
Model PR05502SP
Quantity 4/Pk.
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.