Learn More
Bellco Glass, Inc. PUREGRIP Glass Bottles, Media, Round, Amber, GL45 VersaCap , 100mL
SDP
Catalog No. 502550108
Encompass_Preferred
PUREGRIP Glass Reagent Bottles are chemically inert and stable. When fitted with a plastic pouring ring, dripping can be totally eliminated. As there is only one size of screw thread for all the bottles from 100 mL onwards, the screw caps and pouring rings are fully interchangeable. The bottles, pouring rings and caps are autoclavable and sterilizable.SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 100mLCap Size: GL45 Cap Material: PP (polypropylene) Highest cap temperature rating in the industry, at 142°CColor: AmberOverall Dimension: 56mm x 103mmGraduation Color: White EnamelClass/Quality Grade: Type 1, Class A
Catalog No. 50-255-0108 Supplier Bellco Glass, Inc. Supplier No. 452800654
PUREGRIP Glass Reagent Bottles are chemically inert and stable. When fitted with a plastic pouring ring, dripping can be totally eliminated. As there is only one size of screw thread for all the bottles from 100 mL onwards, the screw caps and pouring rings are fully interchangeable. The bottles, pouring rings and caps are autoclavable and sterilizable.SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 100mLCap Size: GL45 Cap Material: PP (polypropylene) Highest cap temperature rating in the industry, at 142°CColor: AmberOverall Dimension: 56mm x 103mmGraduation Color: White EnamelClass/Quality Grade: Type 1, Class A