Learn More
Bellco Glass, Inc. PUREGRIP Glass Bottles, Media, Round, Amber, GL45 VersaCap , 20,000mL
SDP
Supplier:  Bellco Glass, Inc. 452800662
Encompass_Preferred
PUREGRIP Glass Reagent Bottles are chemically inert and stable. When fitted with a plastic pouring ring, dripping can be totally eliminated. As there is only one size of screw thread for all the bottles from 20000mL onwards, the screw caps and pouring rings are fully interchangeable. The bottles, pouring rings and caps are autoclavable and sterilizable.SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 20000mLCap Size: GL45 Cap Material: PP (polypropylene) Highest cap temperature rating in the industry, at 142°CColor: AmberOverall Dimension: 296mm x 503mmGraduation Color: White EnamelClass/Quality Grade: Type 1, Class A
Catalog No. 50-255-0100