Learn More
Bellco Glass, Inc. PUREGRIP Glass Bottles, Media, Round, Amber, GL45 VersaCap , 10,000mL
SDP
Catalog No. 502550101
Encompass_Preferred
PUREGRIP Glass Reagent Bottles are chemically inert and stable. When fitted with a plastic pouring ring, dripping can be totally eliminated. As there is only one size of screw thread for all the bottles from 10L onwards, the screw caps and pouring rings are fully interchangeable. The bottles, pouring rings, and caps are autoclavable and sterilizable.SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 10LCap Size: GL45Cap Material: PP (polypropylene) Highest cap temperature rating in the industry, at 142°CColor: AmberOverall Dimension: 235mm x 408mmGraduation Color: White EnamelClass/Quality Grade: Type 1, Class A
Catalog No. 50-255-0101 Supplier Bellco Glass, Inc. Supplier No. 452800661
PUREGRIP Glass Reagent Bottles are chemically inert and stable. When fitted with a plastic pouring ring, dripping can be totally eliminated. As there is only one size of screw thread for all the bottles from 10L onwards, the screw caps and pouring rings are fully interchangeable. The bottles, pouring rings, and caps are autoclavable and sterilizable.SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 10LCap Size: GL45Cap Material: PP (polypropylene) Highest cap temperature rating in the industry, at 142°CColor: AmberOverall Dimension: 235mm x 408mmGraduation Color: White EnamelClass/Quality Grade: Type 1, Class A