Learn More
Bellco Glass, Inc. PUREGRIP Glass Carboys, Round, Clear, 83mm VersaCap , 20,000mL, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800670
Encompass_Preferred
PUREGRIP Carboys which are chemically inert, mechanically strong and stable. Complies with ISO 4796-1. Comes with retrace code and white enameled area for marking.SpecificationsMaterial: 3.3 Borosilicate GlassVolume: 20 LCap Size: 83 mmCap Material: PP (polypropylene) may be autoclaved at 15 PSIColor: ClearGraduation Color: Ceramic WhiteQuantity per Case: 1Class/ Quality Grade: USP Type I, Class ANeck Type: ThreadedAutoclavable: Yes
Catalog No. 50-255-0092