Learn More
Bellco Glass, Inc. PUREGRIP Glass Flasks, Erlenmeyer, Clr, GL45 VersaCap , 1,000mL
SDP
Supplier:  Bellco Glass, Inc. 452800665
Encompass_Preferred
PUREGRIP flasks are with Uniform wall thickness making them ideal for heating and mixing of liquids. These flasks come with a neutral pouring ring from capacities of 100mL up to 5L.SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 1LCap Size: GL45 Cap Material: PP (polypropylene) Highest cap temperature rating in the industry, at 142°CColor: ClearOverall Dimension: 131mm x 220mmClass/Quality Grade: Type 1, Class A
Catalog No. 50-255-0097