Learn More
Bellco Glass, Inc. PUREGRIP Glass Flasks, Erlenmeyer, Clr, GL45 VersaCap , 250mL
SDP
Supplier:  Bellco Glass, Inc. 452800663
Encompass_Preferred
PUREGRIP flasks are with Uniform wall thickness making them ideal for heating and mixing of liquids. These flasks come with neutral pouring ring from capacities of 100 ml up to 5 Litre. SpecificationsMaterial: 3.3 Borosilicate Glass Bottle Volume: 250mLCap Size: GL45 Cap Material: PP (polypropylene) Highest cap temperature rating in the industry, at 142°CColor: ClearOverall Dimension: 85mm x 145mmClass/Quality Grade: Type 1, Class A
Catalog No. 50-255-0099