Learn More
Bellco Glass, Inc. PUREGRIP Universal  HPLC Solvent Reservoir  Bottle Assembly, VersaCap GL45, 500mL Clr Reagent Bottle,  2 X Ports- OD Tube 3.2mm (1/8") & 1.6mm (1/16"), 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800084
Encompass_Preferred
PUREGRIP Universal  HPLC Solvent Reservoir  Bottle Assembly, VersaCap GL45, 500mL Clr Reagent Bottle,  2 X Ports- OD Tube 3.2mm (1/8") & 1.6mm (1/16"), 1/EA
Catalog No. 50-255-0678