Learn More
Bellco Glass, Inc. PUREGRIP Universal HPLC Solvent Reservoir  Bottle Assembly, VersaCap GL45, 1L Clr Reagent Bottle,  2X Ports- OD Tube 3.2mm (1/8") & 1.6mm (1/16"), 1/EA
SDP
Catalog No. 502550673
Encompass_Preferred
PUREGRIP Universal HPLC Solvent Reservoir  Bottle Assembly, VersaCap GL45, 1L Clr Reagent Bottle,  2X Ports- OD Tube 3.2mm (1/8") & 1.6mm (1/16"), 1/EA
Catalog No. 50-255-0673 Supplier Bellco Glass, Inc. Supplier No. 452800089
PUREGRIP Universal HPLC Solvent Reservoir  Bottle Assembly, VersaCap GL45, 1L Clr Reagent Bottle,  2X Ports- OD Tube 3.2mm (1/8") & 1.6mm (1/16"), 1/EA