Learn More
Braintree Scientific Pureline? Oxygen Concentrator Filter, intake filter
SDP
Supplier:  Braintree Scientific OC40002
Encompass
INTAKE FILTER - CHANGE FINE INTAKE FILTERS DURING YEARLY ANESTHESIA SERVICE CALL OR EVERY 2 YEARS. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5423