Learn More
Braintree Scientific Pureline? Oxygen Concentrator Filter, intake filter
SDP
Catalog No. 501955426
Encompass
CHANGE FINE INTAKE FILTERS DURING YEARLY ANESTHESIA SERVICE CALL OR EVERY 2 YEARS. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5426 Supplier Braintree Scientific Supplier No. OC80002
CHANGE FINE INTAKE FILTERS DURING YEARLY ANESTHESIA SERVICE CALL OR EVERY 2 YEARS. **Sold for research purposes only and not for human use**