Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ Genomic DNA Mini Kit

Catalog No. k182001
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 Preps
250 Preps
50 Preps
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K182001 50 Preps
K182000 10 Preps
K182002 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. K182001 Supplier Invitrogen™ Supplier No. K182001
Only null left
Add to Cart
Edge
Add to Cart

The PureLink™ Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format.

The PureLink™ Genomic DNA Mini Kit enables high-yield, high-purity, genomic DNA (gDNA) extractions from a wide variety of sample types. Use this single kit for genomic DNA purification from blood, tissues, cells, bacteria, swabs, and blood spots, with a familiar silica-based, microcentrifuge spin-column format. Features of the PureLink™ Genomic DNA Mini Kit include:

Kit flexibility—one kit works with a variety of sample types and sizes
Ultraclean gDNA—minimal contamination of DNA product means successful downstream application
Improved design—optimized spin-column design and buffer formulation for better yield and purity

Multiple sample sources, one kit
The PureLink™ Genomic DNA Mini Kit can be used with many different sample types, each with its own optimized protocol outlined in the manual.

Optimized components and protocols
The PureLink™ Genomic DNA Mini Kit includes parts and protocols that improve upon the easy and familiar spin-column methods currently in use. Columns are newly configured to bind and release DNA more efficiently. Each sample source uses a specialized lysis procedure and an optimized lysis buffer formulated to enhance proteinase K activity and eliminate protein contamination. A chaotropic salt buffer promotes stable binding of DNA to the column resin, and powerful wash buffers remove all traces of protein and salt. DNA is eluted in a low-salt buffer to allow for pH stabilization of the DNA in storage.

Higher yields and purity
With PureLink™ Genomic DNA Kits you can expect high yields of high-purity gDNA (determined by A260/A280 measurements), no matter the sample type, from bacteria to tissues to blood to cultured cells. The enhanced PureLink™ Lysis and Wash Buffers help to consistently deliver purified gDNA samples with absorbance reading ratios of ∼1.9 for both A260/A280 and A260/A230, indicating no detectable contamination from carryover of protein or guanidine.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 10 mL PureLink™ Genomic Lysis/Binding Buffer
• 9 mL PureLink™ Genomic Digestion Buffer
• 10 mL PureLink™ Genomic Wash Buffer 1
• 7.5 mL PureLink™ Genomic Wash Buffer 2
• 10 mL PureLink™ Genomic Elution Buffer
• 1 mL RNase A (20 mg/mL)
• 1 mL Proteinase K (20 mg/mL)
• 50 each PureLink™ Spin Columns with Collection Tubes
• 100 PureLink™ Collecion Tubes (2.0 mL)

This kit contains enough reagents for 50 DNA preps. Store all components at room temperature. For long-term storage, Proteinase K and RNase A may be stored at 4°C.
Isolation Technology Silica Spin Column
Scale Mini
Sample Type Bacteria, Blood, Cells, Tissue
For Use With (Application) Genotyping, Real-Time Quantitative PCR (qPCR), Southern Blotting, Sequencing, PCR
High-throughput Compatibility Not High-throughput Compatible (Manual)
Prep Scale 100 μL to 1 mL Blood Samples (Medium-Scale)
Product Line PureLink
Product Type Genomic DNA Mini Kit
Quantity 50 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.