Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PureLink™ HiPure Expi Plasmid Gigaprep Kit

Catalog No. K210009XP
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K210009XP 2 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K210009XP Supplier Invitrogen™ Supplier No. K210009XP
  • PRODUCT PROMOTION
    $333.00
    $281.00 / Each of 1

    Save $52.00

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The PureLink™ HiPure Plasmid Plasmid Purification Kits also include vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times.

The PureLink™ HiPure Plasmid Plasmid Purification Kits also include vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times.

The PureLink™ HiPure Expi Plasmid Plasmid Purification Kits enable isolation of the highest levels of transfection-grade plasmid DNA from a single E. coli prep using an enhanced anion exchange resin. The PureLink™ HiPure Plasmid Plasmid Purification Kits also include vacuum filtration columns that provide bacterial filtration without centrifugation, enabling extremely fast process times. The PureLink™ HiPure Expi Plasmid Plasmid Purification Kits typically isolate 12 mg of high quality, ultrapure plasmid DNA with inherently low endotoxin levels from 2.5 L of bacterial culture.

Advantages of the PureLink™ HiPure Expi Plasmid Plasmid Purification Kits include:

  • High yield—isolate up to 14 mg of high quality plasmid DNA from a single purification using 5 L of bacterial culture volume
  • Purity—inherently low endotoxin levels (typically 0.1–1.0 EU/μg) and A260/280 >1.8, making it ideal for mammalian cell transfection
  • Simple and super-fast protocol—with vacuum assisted protocol, typical plasmid isolation in as little as 90 minutes

HiPure Expi anion exchange chromatography for plasmid purification

PureLink™ HiPure Expi plasmid purification kits use a novel, proprietary anion-exchange resin to enable the purification of the highest levels of transfection-grade plasmid DNA. The enhanced resin combines large capacity with fast flow rate, high resolution, and efficient endotoxin removal. Plasmid preparations are typically completed in less than 2 hours with purity equal to two CsCl gradients. PureLink™ HiPure Expi plasmid purification kits enable the quickest process for obtaining the highest amount of transfection-grade plasmid DNA.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Equilibration Buffer
• Resuspension Buffer
• Lysis Buffer
• Precipitation Buffer
• RNase A
• Wash Buffer
• Elution Buffer
• TE Buffer
• Expi HiPure Giga Filtration Column
• Expi HiPure Giga DNA Binding Column

Store all components at room temperature upon receipt.
Format Kit
Isolation Technology Anion Exchange Resin
Sample Type Bacterial Culture
For Use With (Application) Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription
High-throughput Compatibility Manual Protocols
Prep Scale 12 mg plasmid DNA
Product Line PureLink
Product Type Plasmid GigaPrep Kit
Quantity 2 Preps
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Target Plasmid DNA
Test Time 2 hr.
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.