Learn More
Invitrogen™ PureLink™ Quick Gel Extraction Kit

Description
Includes
Gel Solubilization Buffer (L3)
Wash Buffer (W1)
Elution Buffer (E5); (10 mM Tris-HCl, pH 8.5)
Quick Gel Extraction Columns
Wash Tubes
Recovery Tubes
The PureLink™ Quick Gel Extraction Kit allows you to rapidly and efficiently purify DNA fragments from TAE or TBE agarose gels of various percentages. DNA can be extracted and purified from agarose gels with different melting points in ∼30 minutes using PureLink™ silica membrane-based quick gel extraction columns. For your convenience, purification protocols are provided for centrifugation and for vacuum.
Advantages of using PureLink™ Quick Gel Extraction Kit:
- Purify DNA fragments from TAE and TBE agarose gels of various percentages and melting points
- Complete the procedure in ∼30 minutes
- Easily purify DNA fragments from 40 bp to 10 kb from gels
- Obtain high recovery of DNA fragments
- Bind and purify up to 15 μg DNA with one column
- Purify DNA fragments that are high quality and show reliable performance in PCR, restriction enzyme digestion, cloning, and labeling
Note: The PureLink™ Quick Gel Extraction Kit is not designed to purify supercoiled plasmid DNA or genomic DNA from agarose gels. Only linear DNA fragments may be purified from gels using this kit.
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | All components of the PureLink™ Quick Gel Extraction System are shipped at room temperature. Upon receipt, store all components at room temperature. |
| Format | Kit |
| Isolation Technology | Spin or Vacuum Column, Silica Spin Column |
| Sample Type | DNA Fragments in Agarose Gel Slices, Gel Samples |
| For Use With (Application) | Next-Generation Sequencing, Cloning, Sequencing, Nucleic Acid Labeling, PCR, In Vitro Transcription |
| High-throughput Compatibility | Not High-throughput Compatible (Manual) |
| Label or Dye | Ethidium Bromide, SYBR Safe |
| Product Line | PureLink |
| Product Type | Quick Gel Extraction Kit |
| Quantity | 250 Preps |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.