Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Advanced Hand Sanitizer E3 Rated Foam
GREENER_CHOICESDP

Catalog No. 19044048 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-044-048 1200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-044-048 Supplier Gojo Industries Inc Supplier No. 539302
Add to Cart
Add to Cart
This item is not returnable. View return policy

E3 rated foam sanitizer refill for TFX™ touch-free dispenser.

  • Fragrance free foam certified for use in food processing facilities
  • Kills 99.99 percent of common germs that may cause illness
  • Listed with NSF International as an E3 for use in all departments of USDA regulated food processing environments
  • ECOLOGO™ Certified to Standard Name UL 2783
  • Approved for hand/food contact after handwashing
  • Certified Kosher
  • Outperforms other hand sanitizers ounce for ounce
  • Clinically-proven to maintain skin health
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
Form Foam
Description Advanced Hand Sanitizer E3 Rated Foam Refill
For Use With (Equipment) Purell TFX Dispensers
Size 1.2 L
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.