Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Advanced Hand Sanitizer Gel: Refill for NXT™ Dispenser
SDP

Catalog No. 19043910 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
2000 mL
Quantity:
4 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity
19-043-910 2000 mL 4 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-910 Supplier Gojo Industries Inc Supplier No. 225604

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Refill for use with Purell™ NXT™ Dispensers.

  • Kills of most common germs that may cause illness
  • Maintains skin health
  • Refill pack options feature removable pump for easier recycling and are SANITARY SEALED™ to prevent contamination
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Form Gel
For Use With (Equipment) Refill for PURELL™ NXT™ Dispenser
Quantity 4 Pk.
Volume (Metric) 2000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.