Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Advanced Hand Sanitizer Green Certified Gel
GREENER_CHOICESDP

Catalog No. 19043917 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Quantity:
12/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-043-917 12/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-917 Supplier Gojo Industries Inc Supplier No. 369112
  • $164.30
    $149.65 / Case of 12

    Save $14.65

    9% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Refreshing gel made with naturally-renewable ethanol.

  • Kills most common germs that may cause illness
  • Maintains skin health
  • ECOLOGO™ Certified to Standard Name UL 2783
  • USDA Certified Biobased Product
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
Form Gel
Certifications/Compliance Green Certified
For Use With (Equipment) Table Top Pump Bottle
Quantity 12/Pk.
Size 12 oz.
Volume (English) 12 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.