Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GOJO™ Purell™ Body Fluid Spill Kit
SDP

Catalog No. 19043880 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Product Type:
Body Fluid Spill Kit
Body Fluid Spill Kit Refill
Packaging:
Clear Plastic Bag
Hard Plastic Clamshell
White Plastic Bag
Dimensions (L x W):
22 x 14 in.
22 x 16 in.
27.5 x 17.5 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Packaging Dimensions (L x W)
19-043-880 Body Fluid Spill Kit Hard Plastic Clamshell 22 x 16 in.
19-043-878 Body Fluid Spill Kit Refill Clear Plastic Bag 22 x 14 in.
19-043-879 Body Fluid Spill Kit White Plastic Bag 27.5 x 17.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-043-880 Supplier GOJO™ Supplier No. 384108CLMS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Body fluid spill kit features head-to-toe personal protection and cleanup products to safely remove body fluid spills, and help prevent the spread of germs that can cause illness.

  • Provides a comprehensive solution to help reduce the risk of Norovirus outbreaks
  • Safely removes vomit, blood, feces & other body fluids
  • Effective against Norovirus, E.Coli, Salmonella and more in just 30 seconds
  • Proven effective on most hard and soft surfaces
  • Protects staff and patrons from exposure to biohazards
  • Bilingual, step-by-step instructions for ease of use
  • OSHA and Food Code compliant
  • Approved for food contact surfaces (no rinse required)
  • Includes hairnet, sleeved gown, gloves, shoe covers, eye shield and mask to provide head-to-toe personal protection
  • 3-year shelf life

Compliance

OSHA and Food Code Compliant

TRUSTED_SUSTAINABILITY

Specifications

Product Type Body Fluid Spill Kit
For Use With (Application) Body fluid spills
Length (English) 22 in.
Length (Metric) 55.88 cm
Color Clear
Material Plastic
Sterility Non-sterile
Packaging Hard Plastic Clamshell
Quantity 8/Pk.
Dimensions (L x W) 22 x 16 in.
Width (English) 16 in
Width (Metric) 40.64 cm
Height (Metric) 30.48 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.