Learn More
GOJO PURELL™ DS360 High-Capacity Surface Disinfecting Wipes Station
SDP
Catalog No. 17100924 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Holds:
Wipes
Catalog No. Holds
17-100-924 Wipes
1 options
Catalog No. 17-100-924 Supplier GOJO Supplier No. 911701SLVSFWP
Add to Cart
Add to Cart
360 - Degree access wipes dispenser with rust-resistant design, waste receptacle, and storage compartment for clean, organized facility hygiene.
  • Premium high-capacity floor stand dispenser for quick and convenient surface hygiene.
  • Features the REDIPUL™ Nozzle that virtually eliminates wipe fallback, ensuring a single wipe is always ready.
  • Patented re-wicking technology prevents wipes from drying out for consistent performance.
  • Durable steel frame construction optimized to prevent rusting for long-lasting reliability.
  • Freestanding weighted base provides stability and prevents movement during use.
  • Integrated waste receptacle supports cleanliness and organization in high-traffic areas.
  • Built-in compartment allows storage of additional wipe refills for uninterrupted use.
  • Compatible with PURELL™ Surface Multi-Purpose Disinfecting Wipes, 1200 Count Refill (9103-02), sold separately.
  • Includes one PURELL™ DS360 High-Capacity Surface Disinfecting Wipes Station, Silver.
  • Proud to be certified as a Women's Business Enterprise by the Women's Business Enterprise National Council (WBENC).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wiper Dispenser
Color Graphite
Holds Wipes
Material Metal
Mounting Freestanding
No. of Compartments 3
Product Style Front-loading with Opening
Sterility Non-sterile