Learn More
PURELL™ Hand Sanitizing Wipes, High Capacity Dispenser Refill
Shop All Gojo Industries, Inc. Products

Description
- Large, durable, and textured wipe removes light soils and dirt from hands
- Kills most common germs that may cause illness
- Convenient for use whenever soap and water are not available
- Dermatologist tested
- Alcohol-free formulation
Specifications
Specifications
| Product Type | Sanitizing Wipe |
| Form | Wipe |
| For Use With (Equipment) | PURELL™ High Capacity Wipes Dispensers |
| Quantity | 1700 count |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.