Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Hand Sanitizing Wipes High Capacity Floor Stand Dispenser
SDP

Catalog No. 19043940 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Color:
Black
Blue
Silver
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-043-940 Blue
19-043-941 Black
19-043-942 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-043-940 Supplier Gojo Industries Inc Supplier No. 9118DS

High capacity floor stand dispenser for PURELL™ wipes

  • Weighted base keeps stand in place
  • Durable, freestanding unit
  • Pre-installed SIMPUL™ nozzle technology, virtually eliminates roping, providing one wipe every time
  • Storage compartment fits 6,000 wipes
  • Available with customizable signage
  • Available in blue, black and reflective silver
TRUSTED_SUSTAINABILITY

Specifications

Product Type Stand Dispenser
Color Blue
Width (English) 15.75 in.
Height (English) 39.16 in.
Capacity 6000 Wipes
For Use With (Equipment) PURELL™ Hand Sanitizing Wipes
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.