Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURELL™ Healthcare Advanced Hand Sanitizer Foam
SDP

Catalog No. 19044037 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
ES4 dispensing system
ES6 dispensing system
ES8 dispensing system
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-044-037 ES4 dispensing system
19-043-952 ES6 dispensing system
19-043-955 ES8 dispensing system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-044-037 Supplier Gojo Industries Inc Supplier No. 505302
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hand sanitizer foam is a refill for touch-free hand sanitizer dispensers.

Advanced Hand Sanitizer Foam formulation designed for healthcare environments. AT-A-GLANCE refills make it obvious when refills need to be changed.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
Form Foam
For Use With (Equipment) ES4 dispensing system
Size 1200 mL
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.