Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PURELL™ Healthcare Advanced Hand Sanitizer Gel
SDP

Catalog No. 19043947 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-043-947 1200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-043-947 Supplier Gojo Industries Inc Supplier No. 506302
Add to Cart
Add to Cart
This item is not returnable. View return policy

Refill for PURELL™ ES4 Push-Style Hand Sanitizer Dispenser.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
Form Gel
Description Citrus scent
For Use With (Application) Hands
Quantity 2/Pk.
Size 1200 mL
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.