Learn More
Description
- Freestanding design, ideal for high-traffic areas
- Makes Purell™ Instant Hand Sanitizer available anywhere
- Dispenser sold separately
Specifications
Specifications
| Product Type | Floor Stand Dispenser |
| Color | Silver |
| For Use With (Application) | High traffic areas, such as reception areas or cafeterias |
| For Use With (Equipment) | Touch-free dispensers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
