Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gojo™ Purell™ LTX™ or TFX™ Dispenser Floor Stand
SDP

Catalog No. 19043949 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Color:
Black
Silver
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-043-949 Black
19-043-948 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-043-949 Supplier Gojo™ Supplier No. 2425DS

Floor stand for use with Purell™ LTX™ or TFX™ dispensers.

  • Freestanding design, ideal for high-traffic areas
  • Makes Purell™ Instant Hand Sanitizer available anywhere
  • Dispenser sold separately
TRUSTED_SUSTAINABILITY

Specifications

Product Type Floor Stand Dispenser
Color Black
For Use With (Application) High traffic areas, such as reception areas or cafeterias
For Use With (Equipment) Touch-free dispensers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.