Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gojo™ Purell™ LTX™ or TFX™ Dispenser Floor Stand
SDP

Catalog No. p-9012000 Shop All Gojo Industries, Inc. Products
Click to view available options
Color:
Black
Silver

Catalog No. 19-043-948

Supplier: Gojo™ 2423DS

Floor stand for use with Purell™ LTX™ or TFX™ dispensers.

  • Freestanding design, ideal for high-traffic areas
  • Makes Purell™ Instant Hand Sanitizer available anywhere
  • Dispenser sold separately
TRUSTED_SUSTAINABILITY

Specifications

Product Type Floor Stand Dispenser
Color Silver
For Use With (Application) High traffic areas, such as reception areas or cafeterias
For Use With (Equipment) Touch-free dispensers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.