Description
- Federal, state and local health regulations require on-site testing of surface sanitizers to verify their effectiveness.
- Active ingredient test strips help your facility remain compliant by confirming the efficacy of PURELL™ Surface Sanitizing and Disinfecting Wipes.
- Quickly and accurately measure the active ingredient level of PURELL™ Surface Sanitizing and Disinfecting Wipes (20% ethyl alcohol)
- Remain in compliance with federal, state, and local health department regulations
- Easy-to-read instructions and images make testing simple
- Includes ten individual test strips and instructions/color matching chart on bottle label
- For use only with PURELL™ Surface Sanitizing and Disinfecting Wipes
Specifications
Specifications
| Product Type | Test Strip |
| Material | Paper |
| Container Type | Bottle |
| For Use With (Application) | PURELL™ Surface Sanitizing and Disinfecting Wipes |