Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ VF PLUS Hand Sanitizer Gel
GREENER_CHOICESDP

Catalog No. 19044049 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Volume (Metric):
1200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-044-049 1200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-044-049 Supplier Gojo Industries Inc Supplier No. 549904
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VF PLUS Hand Sanitizer Gel Refill for TFX™ Touch-free Dispenser.

  • Provides protection for healthcare professionals and patients in real working conditions
  • Broad spectrum of activity
  • Kills up to 99.999% of the most common potentially dangerous germs
  • Fragrance free
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
Form Gel
Description VF PLUS Hand Sanitizer Gel Refill
For Use With (Equipment) Purell TFX Dispensers
Size 1200 mL
Volume (Metric) 1200 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.