Learn More
Thomas Scientific PurePlus 0.2mL 8-Well PCR Tube Strips, 120pk
SDP
Catalog No. NC1732516
Encompass_Preferred
PurePlus 0.2mL 8-Well PCR Tube Strips with Individually Attached Bubble Caps, in Bags. With individually attached bubble snap caps.\ATP, RNase/DNase and Endotoxin Free (non-pyrogenic).Autoclavable
Catalog No. NC1732516 Supplier Thomas Scientific Supplier No. 1141N56
PurePlus 0.2mL 8-Well PCR Tube Strips with Individually Attached Bubble Caps, in Bags. With individually attached bubble snap caps.\ATP, RNase/DNase and Endotoxin Free (non-pyrogenic).Autoclavable