Learn More
Labcon North America Labcon PurePlus Deep Well Plate, 2.0 mL Capacity, Non-Pyrogenic, Autoclavable, 50 per Case
Catalog No. NC3779863
Encompass_Preferred
The Labcon PurePlus Deep Well Plate is a reliable 96-well plate with a 2.0 mL capacity, designed for high purity scientific applications. Made from 100% virgin USP Class VI polypropylene, it is free from RNase, DNase, ATP, and endotoxins. Features include round wells, conical bottoms, a notched registration corner for easy orientation, and a robust design suitable for both manual and robotic handling. Autoclavable and freezable to -80°C, it offers exceptional versatility in laboratory processes.
Catalog No. NC3779863 Supplier Labcon North America Supplier No. 39015200009
The Labcon PurePlus Deep Well Plate is a reliable 96-well plate with a 2.0 mL capacity, designed for high purity scientific applications. Made from 100% virgin USP Class VI polypropylene, it is free from RNase, DNase, ATP, and endotoxins. Features include round wells, conical bottoms, a notched registration corner for easy orientation, and a robust design suitable for both manual and robotic handling. Autoclavable and freezable to -80°C, it offers exceptional versatility in laboratory processes.