Learn More
Alkali Scientific PurePoint™ 10ul XL Reload Refillable Pipette Tips, 960/box, 3840/Case
SDP
Catalog No. 5000663203
Encompass_Preferred
PurePoint™ 10ul XL Reload Refillable Pipette Tips are designed for accuracy in small volume dispensing. With a capacity of 960 tips per box and 3840 tips per case, these extra-long tips offer superior precision and minimal liquid retention. The refillable design ensures cost-effective use, while the low retention feature guarantees more accurate sample handling. Perfect for laboratories that require consistent, reliable pipetting performance.
Catalog No. 50-006-63203 Supplier Alkali Scientific Supplier No. RT1010CS
PurePoint™ 10ul XL Reload Refillable Pipette Tips are designed for accuracy in small volume dispensing. With a capacity of 960 tips per box and 3840 tips per case, these extra-long tips offer superior precision and minimal liquid retention. The refillable design ensures cost-effective use, while the low retention feature guarantees more accurate sample handling. Perfect for laboratories that require consistent, reliable pipetting performance.