Learn More
Alkali Scientific PurePoint™ Rainin LTS™ LiteTouch™ Compatible 250uL Reload Stackable Tips - Low Retention, 960/box, 9600/Case
SDP
Catalog No. 503028269
Encompass_Preferred
PurePoint LTS Pipette Tips fit Rainin LTS Pipettes and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, Pyrogen Free, and range in volume from 20µL to 1000µL. The environmentally friendly packaging allows you to reuse most racks and produces up to 75% less plastic waste. Stackable design reduces storage Compatibile with Rainin LTS™ LiteTouch™ and XLS+ Pipettes Environmentally friendly — 75% less waste Great for multichannels Low forces — reduces tip ejection force up to 85% Low retention — most precise, accurate delivery RNAse, DNAse, Pyrogen Free Sterile
Catalog No. 50-302-8269 Supplier Alkali Scientific Supplier No. RLT250CS
PurePoint LTS Pipette Tips fit Rainin LTS Pipettes and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, Pyrogen Free, and range in volume from 20µL to 1000µL. The environmentally friendly packaging allows you to reuse most racks and produces up to 75% less plastic waste. Stackable design reduces storage Compatibile with Rainin LTS™ LiteTouch™ and XLS+ Pipettes Environmentally friendly — 75% less waste Great for multichannels Low forces — reduces tip ejection force up to 85% Low retention — most precise, accurate delivery RNAse, DNAse, Pyrogen Free Sterile