Learn More
Alkali Scientific PurePoint™ Rainin LTS™ LiteTouch™ Compatible 200uL Filter Pipette Tips - Low Retention, 960/box, 4800/case
SDP
Catalog No. 503028905
Encompass_Preferred
PurePoint LTS Filter Tips fit Rainin* LTS and XLS Pipettes, and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, Pyrogen Free, and range in volume from 20µL to 1000µL. Low forces — reduces tip ejection force up to 85% Low retention — most precise, accurate delivery Stable new rack designed for multichannel pipettors Compatibile Rainin LTS™ LiteTouch™ and XLS+ Pipettes RNAse, DNAse, Pyrogen Free Sterile *Lite Touch™ is a registered trademark of Mettler-Toledo Rainin, LLC
Catalog No. 50-302-8905 Supplier Alkali Scientific Supplier No. FLT200CS
PurePoint LTS Filter Tips fit Rainin* LTS and XLS Pipettes, and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, Pyrogen Free, and range in volume from 20µL to 1000µL. Low forces — reduces tip ejection force up to 85% Low retention — most precise, accurate delivery Stable new rack designed for multichannel pipettors Compatibile Rainin LTS™ LiteTouch™ and XLS+ Pipettes RNAse, DNAse, Pyrogen Free Sterile *Lite Touch™ is a registered trademark of Mettler-Toledo Rainin, LLC