Learn More
Alkali Scientific PurePoint™ Sterile Tips Racked - Low Retention, 20uL, 960/Pk
SDP
Catalog No. 503029075
Encompass_Preferred
Our PurePoint™ Sterile Racked LTS Tips fit Rainin LTS Pipettes and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, Pyrogen Free, and range in volume from 20µL to 1000µL. Low forces — reduces tip ejection force up to 85% Low retention — most precise, accurate delivery Stable new rack designed for multichannel pipettors Compatibile with Rainin LTS™ LiteTouch™ and XLS+ Pipettes RNAse, DNAse, Pyrogen Free Sterile
Catalog No. 50-302-9075 Supplier Alkali Scientific Supplier No. SLT20N
Our PurePoint™ Sterile Racked LTS Tips fit Rainin LTS Pipettes and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, Pyrogen Free, and range in volume from 20µL to 1000µL. Low forces — reduces tip ejection force up to 85% Low retention — most precise, accurate delivery Stable new rack designed for multichannel pipettors Compatibile with Rainin LTS™ LiteTouch™ and XLS+ Pipettes RNAse, DNAse, Pyrogen Free Sterile