Learn More
Alkali Scientific PurePoint™ 5mL Filter Pipet Tips, Sterile, 30 Racks of 24, 720/cs
SDP
Catalog No. 502335951
Encompass_Preferred
A superior fit for your universal fit pipettes such as Gilson and Eppendorf. PurePoint™ tips to provide a perfect fit and deliver reliable performance every time. Offer improved accuracy over serological pipets. The tip length allows better access to the bottom of test tubes, reagent bottles, flasks and other vessels. These 5mL macro tips are graduated at 0.1mL increments. Tips are manufactured from 100% virgin polypropylene and should not be autoclaved, they are sterile upon arrival. PurePoint™ Tips are Certified RNase/DNase free and non-pyrogenic. 720 tips per case.
Catalog No. 50-233-5951 Supplier Alkali Scientific Supplier No. FT95ML
A superior fit for your universal fit pipettes such as Gilson and Eppendorf. PurePoint™ tips to provide a perfect fit and deliver reliable performance every time. Offer improved accuracy over serological pipets. The tip length allows better access to the bottom of test tubes, reagent bottles, flasks and other vessels. These 5mL macro tips are graduated at 0.1mL increments. Tips are manufactured from 100% virgin polypropylene and should not be autoclaved, they are sterile upon arrival. PurePoint™ Tips are Certified RNase/DNase free and non-pyrogenic. 720 tips per case.