Learn More
Alkali Scientific PurePoint™ LTS Filter Tips, 1000µL Max Volume for Rainin 1000μL Pipettes with LTS™ LiteTouch™, Low Retention, 96 tips/rack, 10 racks, 960 tips/ea
SDP
Catalog No. 502336071
Encompass_Preferred
PurePoint™ LTS Filter Tips fit Rainin* LTS Pipettes, and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, and pyrogen free. Low forces — reduces tip ejection force up to 85%. Low retention — most precise, accurate delivery. Stable new rack designed for multichannel pipettors. Great fit on Rainin* LiteTouchª pipettors. RNAse, DNAse, pyrogen free. Sterile.
Catalog No. 50-233-6071 Supplier Alkali Scientific Supplier No. FLT1000
PurePoint™ LTS Filter Tips fit Rainin* LTS Pipettes, and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, and pyrogen free. Low forces — reduces tip ejection force up to 85%. Low retention — most precise, accurate delivery. Stable new rack designed for multichannel pipettors. Great fit on Rainin* LiteTouchª pipettors. RNAse, DNAse, pyrogen free. Sterile.