Learn More
Alkali Scientific PurePoint™ LTS Tips, 1000 uL, Refills, 96 Tips/Reload, 10 Reloads/pk, 10 pk/cs, 960 Tips/cs
SDP
Catalog No. 502336155
Encompass_Preferred
Environmentally friendly — 75% less waste. Great for multichannels. Low forces — reduces tip ejection force up to 85%. Low retention — most precise, accurate delivery. Great fit on Rainin* LiteTouch pipettors, RNAse, DNAse, pyrogen free, sterile. PurePoint LTS Refill Tips fit Rainin LTS Pipettes and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, pyrogen free, 1000µL. The environmentally friendly packaging allows you to reuse most racks and produces up to 75% less plastic waste.
Catalog No. 50-233-6155 Supplier Alkali Scientific Supplier No. RLT1000
Environmentally friendly — 75% less waste. Great for multichannels. Low forces — reduces tip ejection force up to 85%. Low retention — most precise, accurate delivery. Great fit on Rainin* LiteTouch pipettors, RNAse, DNAse, pyrogen free, sterile. PurePoint LTS Refill Tips fit Rainin LTS Pipettes and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, pyrogen free, 1000µL. The environmentally friendly packaging allows you to reuse most racks and produces up to 75% less plastic waste.