Learn More
Alkali Scientific PurePoint™ LTS Tips, 1000 uL, Sterile, Racked, 96 Tips/Rack, 6 Racks/pk, 576/cs
SDP
Catalog No. 502336094
Encompass_Preferred
PurePoint™ Sterile Racked LTS Tips fit Rainin LTS Pipettes and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, pyrogen free.
Catalog No. 50-233-6094 Supplier Alkali Scientific Supplier No. SLT1000
PurePoint™ Sterile Racked LTS Tips fit Rainin LTS Pipettes and are made of premium-grade low retention resin that enables the most accurate delivery of liquids. They also eject with up to 85% less force than conventional pipette tips and may help reduce repetitive pipetting injuries. They are RNAse, DNAse, pyrogen free.