Learn More
Description
Requires
Magna Grip™ Rack, eight-well (Mfr. No. 20-400)
- Bind 1.5 to 3.5μg of Rabbit IgG per μL of suspension
- More than 6x the binding capacity of competitive magnetic beads
- Total removal of buffers with no sample loss for consistent results
- Beads are immobilized in seconds
- Economical
- Particle size: 10μm
- Not recommended for Chromatin Immunoprecipitation (ChIP)
Serum depletion and immunoprecipitation assays
Specifications
Specifications
| Binding Property | 2.5 to 3.5μg/μl |
| Content And Storage | 2°C to 8°C |
| Particle Size | 10 μm |
| Quantity | 2 x 1 mL |
| For Use With (Application) | Serum depletion, immunoprecipitation |
| Type | Protein G Magnetic Bead System |
Unless otherwise stated in the catalog or other company documentation accompanying the product(s), these products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.