Learn More
Zymo Research Corporation PureRec RNase A
SDP
Catalog No. 502865898
Encompass_Preferred
RNase A is a very active endoribonuclease that digests RNA in a wide range of buffer conditions and temperatures. PureRec RNase A is a recombinant RNase A, overexpressed and extensively purified from a prokaryotic host. It is extremely active, free of DNase and guaranteed animal-free. It is ready to use, suppliedat 1mg/ml concentration.
Catalog No. 50-286-5898 Supplier Zymo Research Corporation Supplier No. E10195
RNase A is a very active endoribonuclease that digests RNA in a wide range of buffer conditions and temperatures. PureRec RNase A is a recombinant RNase A, overexpressed and extensively purified from a prokaryotic host. It is extremely active, free of DNase and guaranteed animal-free. It is ready to use, suppliedat 1mg/ml concentration.