Learn More
Cole-Parmer Chromatography Research Supplies Puresep-T 230685 Septa, 7/16" (11.0 mm); 50/pack
Catalog No. NC1522945 Shop All Cole-Parmer Products
Encompass_Preferred
PURESEP-T™ septa are made of PTFE and polyimide films on silicone rubber bases. PTFE gives maximum inertness and low bleed. Maximum temperature: 437°F (225°C).
Catalog No. NC1522945 Supplier Cole-Parmer Supplier No. 0199422
PURESEP-T™ septa are made of PTFE and polyimide films on silicone rubber bases. PTFE gives maximum inertness and low bleed. Maximum temperature: 437°F (225°C).