Learn More
Sigma Aldrich Fine Chemicals Biosciences PureSolvTM Micro 100 Liter1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z5680311EA
Encompass
Easy access to smaller volumes of anhydrous solventThe Pure Solv Micro solvent purification system is designed as both an economic and space friendly alternative to the larger more traditional free standing units only 12 in. W x 13 in. D x 19 in. H. This purification system will remove water from approximately 100 L of solvent using the same proven design as the Innovative Technology Inc. Pure Solv MD system. Pure Solv Micro is perfect for the researcher with modest requirements for a single anhydrous solvent who prefers the convenience of having a dedicated solvent drying and dispensing station. No longer do you have to share now you can have your own..nbspPure Solv Micro has been packaged so that it can be placed on a bench top inside a fume hood or flammable cabinet as desired by the researcher in charge.nbspPure Solv Micro facilitates proper air free dispensing techniques using an integrated gas and vacuum manifold system. A KNF Laboport solid PTFE vacuum pump 20 L pumping spee
Catalog No. 50-290-9028
Add to Cart
Request a Quote